missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Butyl Sepharose™ 4 Fast Flow Chromatography Media

Catalog No. p-4592274
Change view
Click to view available options
Quantity:
25 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002944 25 mL
45002943 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002944 Supplier Cytiva Supplier No. 17098010
Only null left

Used for capture and intermediate purification of larger proteins

  • Based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 4% agarose
  • Aliphatic butyl ligand immobilized to base matrix with ether linkage
  • Fast Flow matrix gives the adsorbent high chemical, physical and thermal stabilities
  • Dynamic capacity at low salt concentration
  • Used with all aqueous buffers commonly used in chromatography
  • Autoclavable at 121°C for 20 minutes in water
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 25 mL
For Use With (Application) For Capture and Intermediate Purification of Larger Proteins
Product Type Butyl Sepharose 4 Fast Flow Media
Matrix Based on cross-linked 4% agarose