missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Butyl-S Sepharose™ 6 Fast Flow Chromatography Media

Catalog No. 45002941
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002941 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002941 Supplier Cytiva Supplier No. 17097802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The least hydrophobic medium in the Sepharose Fast Flow hydrophobic interaction chromatography (HIC) range and is particularly useful during the initial stages of purification process

  • Binds and elutes relatively strong hydrophobic molecules at comparatively low salt concentrations
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Purifies recombinant Hepatitis B virus surface antigen from CHO cells
  • Composed of cross-linked 6% agarose beads modified with butyl groups via thio-ether linkage
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 200 mL
For Use With (Application) For Hydrophobic Medium in The Sepharose Fast Flow Hydrophobic Interaction Chromatography (HIC) Range and is Particularly Useful During The Initial Stages of PurifiCatIon Process
Stability Stable in commonly used aqueous Buffers: 1 mM HCl, 1 M NaOH, 30% Isopropanol, 50% Ethylene Glycol, 70% Ethanol, 6 M Guanidine hydrochloride, 8 M Urea
Storage Requirements 4°C to 30°C
Product Type Butyl-S Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 20% Ethanol
Ligand Type Butyl-S
Show More Show Less