missing translation for 'onlineSavingsMsg'
Learn More

Butyl Phenyl Ether 99.0+%, TCI America™

Catalog No. p-7118749
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B092725ML 25 mL
B0927500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B092725ML Supplier TCI America Supplier No. B092725ML
Only null left

Chemical Identifiers

CAS 1126-79-0
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00009438
InChI Key YFNONBGXNFCTMM-UHFFFAOYSA-N
Synonym butyl phenyl ether, n-butyl phenyl ether, benzene, butoxy, ether, butyl phenyl, phenyl butyl ether, butoxyphenyl, n-butoxybenzene, butyloxy benzene, n-butylphenylether, butoxy-benzene
PubChem CID 14311
IUPAC Name butoxybenzene
SMILES CCCCOC1=CC=CC=C1

Specifications

Melting Point -19°C
Color Yellow
Boiling Point 210°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Phenyl Ether
RTECSNumber KN5300000
TSCA Yes