missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl n-Octanoate 99.0+%, TCI America™
$96.94 - $96.94
Chemical Identifiers
| CAS | 589-75-3 |
|---|---|
| Molecular Formula | C12H24O2 |
| Molecular Weight (g/mol) | 200.322 |
| MDL Number | MFCD00048918 |
| InChI Key | PSXNDMJWRZYVTM-UHFFFAOYSA-N |
| Synonym | butyl caprylate, butyl n-octanoate, n-butylcaprylate, octanoic acid, butyl ester, n-butyl octanoate, caprylic acid n-butyl ester, n-caprylic acid n-butyl ester, butyl-caprylate, n-butyl n-octanoate, octanoic acid butyl ester |
| PubChem CID | 11517 |
| ChEBI | CHEBI:87380 |
| IUPAC Name | butyl octanoate |
| SMILES | CCCCCCCC(=O)OCCCC |
Chemical Identifiers
| 589-75-3 | |
| 200.322 | |
| PSXNDMJWRZYVTM-UHFFFAOYSA-N | |
| 11517 | |
| butyl octanoate |
| C12H24O2 | |
| MFCD00048918 | |
| butyl caprylate, butyl n-octanoate, n-butylcaprylate, octanoic acid, butyl ester, n-butyl octanoate, caprylic acid n-butyl ester, n-caprylic acid n-butyl ester, butyl-caprylate, n-butyl n-octanoate, octanoic acid butyl ester | |
| CHEBI:87380 | |
| CCCCCCCC(=O)OCCCC |
Specifications
| 589-75-3 | |
| 245°C | |
| MFCD00048918 | |
| butyl caprylate, butyl n-octanoate, n-butylcaprylate, octanoic acid, butyl ester, n-butyl octanoate, caprylic acid n-butyl ester, n-caprylic acid n-butyl ester, butyl-caprylate, n-butyl n-octanoate, octanoic acid butyl ester | |
| CCCCCCCC(=O)OCCCC | |
| 200.322 | |
| CHEBI:87380 | |
| ≥99.0% (GC) | |
| Butyl n-Octanoate |
| Yellow | |
| C12H24O2 | |
| 25 mL | |
| PSXNDMJWRZYVTM-UHFFFAOYSA-N | |
| butyl octanoate | |
| 11517 | |
| 200.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : Yes