accessibility menu, dialog, popup

UserName

CAS RN 589-75-3

H3COOCH3

CAS RN 589-75-3

IUPAC Name: butyl octanoate
Synonyms: butyl octanoate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: PSXNDMJWRZYVTM-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results