missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methyl Ether 99.0+%, TCI America™

$186.74 - $1405.38

Chemical Identifiers

CAS 628-28-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00009453
InChI Key CXBDYQVECUFKRK-UHFFFAOYSA-N
Synonym 1-Methoxybutane
PubChem CID 12338
IUPAC Name 1-methoxybutane
SMILES CCCCOC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M050425ML
View Documents
TCI America
M050425ML
25 mL
Each for $186.74
Only null left
 
M0504100ML
View Documents
TCI America
M0504100ML
100 mL
Each for $283.00
Only null left
 
M0504500ML
View Documents
TCI America
M0504500ML
500 mL
Each for $1,405.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

628-28-4
88.15
CXBDYQVECUFKRK-UHFFFAOYSA-N
12338
CCCCOC
C5H12O
MFCD00009453
1-Methoxybutane
1-methoxybutane

Specifications

628-28-4
70°C
MFCD00009453
2350
CXBDYQVECUFKRK-UHFFFAOYSA-N
1-methoxybutane
12338
≥99.0% (GC)
Butyl Methyl Ether
Colorless
C5H12O
25 mL
1-Methoxybutane
CCCCOC
88.15
88.15
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KN5240000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'