missing translation for 'onlineSavingsMsg'
Learn More

Butyl Methyl Ether 99.0+%, TCI America™

Supplier:  TCI America M0504100ML

 View more versions of this product

Catalog No. M0504100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Butyl Methyl Ether
Colorless
C5H12O
100 mL
1-Methoxybutane
CCCCOC
88.15
88.15
Liquid
628-28-4
70°C
MFCD00009453
2350
CXBDYQVECUFKRK-UHFFFAOYSA-N
1-methoxybutane
12338
≥99.0% (GC)

Chemical Identifiers

628-28-4
88.15
CXBDYQVECUFKRK-UHFFFAOYSA-N
12338
CCCCOC
C5H12O
MFCD00009453
1-Methoxybutane
1-methoxybutane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : KN5240000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions