missing translation for 'onlineSavingsMsg'
Learn More

Butyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7131290
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
250 mL
Catalog No. Quantity
I024925ML 25 mL
I0249250ML 250 mL
2 options

Catalog No. I024925ML

Supplier: TCI America I024925ML

Only null left

Chemical Identifiers

CAS 592-82-5
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004824
InChI Key LIMQQADUEULBSO-UHFFFAOYSA-N
Synonym butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
PubChem CID 11613
ChEBI CHEBI:50534
IUPAC Name 1-isothiocyanatobutane
SMILES CCCCN=C=S

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 2810
Formula Weight 115.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Isothiocyanate
RTECSNumber NX8420500
TSCA Yes