missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Isothiocyanate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 175°C |
| Quantity | 250 mL |
| UN Number | 2810 |
| Formula Weight | 115.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Butyl Isothiocyanate |
Safety and Handling
| RTECSNumber | NX8420500 |
| TSCA | Yes |