missing translation for 'onlineSavingsMsg'
Learn More

Butyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I024925ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I024925ML 25 mL
I0249250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I024925ML Supplier TCI America Supplier No. I024925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 592-82-5
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004824
InChI Key LIMQQADUEULBSO-UHFFFAOYSA-N
Synonym butyl isothiocyanate, n-butyl isothiocyanate, butyl mustard oil, butane, 1-isothiocyanato, 1-butyl isothiocyanate, butylisothiocyanate, isothiocyanic acid, butyl ester, isothiocyanic acid n-butyl ester, 1-isothiocyanato-butane, butanisothiocyanate
PubChem CID 11613
ChEBI CHEBI:50534
IUPAC Name 1-isothiocyanatobutane
SMILES CCCCN=C=S

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 2810
Formula Weight 115.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Isothiocyanate
RTECSNumber NX8420500
TSCA Yes