missing translation for 'onlineSavingsMsg'
Learn More

Butyl Glycidyl Ether 98.0+%, TCI America™

Catalog No. p-7118615
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B069725ML 25 mL
B0697500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B069725ML Supplier TCI America Supplier No. B069725ML
Only null left

Chemical Identifiers

CAS 2426-08-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00005145
InChI Key YSUQLAYJZDEMOT-UHFFFAOYNA-N
Synonym butyl glycidyl ether, n-butyl glycidyl ether, 2-butoxymethyl oxirane, oxirane, butoxymethyl, 1-butoxy-2,3-epoxypropane, glycidyl butyl ether, butoxymethyl oxirane, ether, butyl glycidyl, glycidyl n-butyl ether, butyl 2,3-epoxypropyl ether
PubChem CID 17049
IUPAC Name 2-(butoxymethyl)oxirane
SMILES CCCCOCC1CO1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 3271
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Glycidyl Ether
EINECSNumber (2)-0392
RTECSNumber TX4200000
TSCA Yes