accessibility menu, dialog, popup

UserName

H3COO

CAS RN 2426-08-6

IUPAC Name: 2-(butoxymethyl)oxirane
Synonyms: butyl glycidyl ether N-butyl glycidyl ether glycidyl N-butyl ether
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: YSUQLAYJZDEMOT-UHFFFAOYNA-N
SMILES: CCCCOCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results