missing translation for 'onlineSavingsMsg'
Learn More

Butyl Carbitol™, 20% w/w, Reagents

Catalog No. 01335850
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335850 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335850 Supplier Labchem Inc Supplier No. CB0031001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Butyl Carbitol™, 20% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 112-34-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Colorless
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.99
Percent Purity 20%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Butyl Carbitol™, 20% w/w