missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Benzoate 99.0+%, TCI America™
Supplier: TCI America B0066500ML
Specifications
| Butyl Benzoate | |
| -22°C | |
| 250°C | |
| MFCD00009439 | |
| n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech | |
| CCCCOC(=O)C1=CC=CC=C1 | |
| 178.23 | |
| 178.23 | |
| Liquid |
| 136-60-7 | |
| Colorless | |
| C11H14O2 | |
| 500 mL | |
| XSIFPSYPOVKYCO-UHFFFAOYSA-N | |
| butyl benzoate | |
| 8698 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 136-60-7 | |
| 178.23 | |
| XSIFPSYPOVKYCO-UHFFFAOYSA-N | |
| 8698 | |
| CCCCOC(=O)C1=CC=CC=C1 |
| C11H14O2 | |
| MFCD00009439 | |
| n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech | |
| butyl benzoate |
Safety and Handling
EINECSNumber : (3)-1356
RTECSNumber : DG4925000
TSCA : Yes