missing translation for 'onlineSavingsMsg'
Learn More

Butyl Benzoate 99.0+%, TCI America™

Catalog No. B006625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006625ML 25 mL
B0066500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B006625ML Supplier TCI America Supplier No. B006625ML

Chemical Identifiers

CAS 136-60-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00009439
InChI Key XSIFPSYPOVKYCO-UHFFFAOYSA-N
Synonym n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
PubChem CID 8698
IUPAC Name butyl benzoate
SMILES CCCCOC(=O)C1=CC=CC=C1

Specifications

Melting Point -22°C
Color Colorless
Boiling Point 250°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DG4925000
TSCA Yes