missing translation for 'onlineSavingsMsg'
Learn More

Butyl acetate, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020611
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020611 1 mL
111019298 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020611 Supplier Merck Emd Millipore Supplier No. 732851ML
Only null left
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 790°F
CAS 123-86-4
Vapor Pressure 15 mmHg (25°C)
Vapor Density 4 (vs. air)
Density 0.88 g/mL (at 25°C)
Flash Point 27°C - Closed Cup
Physical Form Neat
Linear Formula CH3 COO(CH2)3 CH3
MDL Number MFCD00009445
Melting Point -78°C
Boiling Point 124°C to 126°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 116.16
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less