missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. p-7130409
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
H021025G 25 g
H0210500G 500 g
2 options

Catalog No. H021025G

Supplier: TCI America H021025G

Only null left

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00016478
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
Synonym butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl
PubChem CID 7184
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 70°C
Color White
Boiling Point 156°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH1980000
TSCA Yes