missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Carboxyphenyl Carbonate 98.0+%, TCI America™

Catalog No. C07731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07731G 1 g
C07735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07731G Supplier TCI America Supplier No. C07731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14180-12-2
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.239
MDL Number MFCD00060074
InChI Key GNBKEARJFHTJMV-UHFFFAOYSA-N
Synonym 4-Carboxyphenyl Butyl Carbonate
PubChem CID 586595
IUPAC Name 4-butoxycarbonyloxybenzoic acid
SMILES CCCCOC(=O)OC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 238.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl 4-Carboxyphenyl Carbonate