accessibility menu, dialog, popup

UserName

H3COOOOOH

CAS RN 14180-12-2

IUPAC Name: 4-[(butoxycarbonyl)oxy]benzoic acid
Synonyms: 4-[(butoxycarbonyl)oxy]benzoic acid
Molecular Weight (g/mol): 238.24
Molecular Formula: C12H14O5
InChi Key: GNBKEARJFHTJMV-UHFFFAOYSA-N
SMILES: CCCCOC(=O)OC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results