missing translation for 'onlineSavingsMsg'
Learn More

Butoxamine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. p-7187586
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014675 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014675 Supplier Merck Emd Millipore Supplier No. B138550MG
Only null left

Butoxamine hydrochloride is a β-andrenoceptor antagonist, which shows β2-selectivity.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5696-15-1
Physical Form Neat
Linear Formula C15H25 NO3 · HCl
MDL Number MFCD00135781
Synonym alpha-(1-[t-Butylamino]ethyl)-2,5-dimethoxybenzyl alcohol
Type Analytical Standard
Industry Type Drug Analysis
Quantity 50 mg
Formula Weight 303.82
Grade Analytical Standard