missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butoxamine hydrochloride, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 5696-15-1 |
| Physical Form | Neat |
| Linear Formula | C15H25 NO3 · HCl |
| MDL Number | MFCD00135781 |
| Synonym | alpha-(1-[t-Butylamino]ethyl)-2,5-dimethoxybenzyl alcohol |
| Type | Analytical Standard |
| Industry Type | Drug Analysis |
| Quantity | 50 mg |
| Formula Weight | 303.82 |
| Grade | Analytical Standard |