missing translation for 'onlineSavingsMsg'
Learn More

Butorphanol tartrate Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. B0371ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0371ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0371ML Supplier MilliporeSigma Supplier No. B0371ML
  • Butorphanol is a synthetic opioid used as a treatment for migraines or moderate to severe pain.
  • This Certified Snap-N-Spike™ Solution is suitable for use as calibrators or controls in a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing to pain prescription monitoring and clinical toxicology.

Specifications

Chemical Name or Material Butorphanol Tartrate
Packaging Ampule
CAS 58786-99-5
Molecular Formula C25H35NO8
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 477.55
Quantity 1 mL
Grade Certified
Show More Show Less