missing translation for 'onlineSavingsMsg'
Learn More

Butoconazole Nitrate 98.0+%, TCI America™

Catalog No. B48011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48011G Supplier TCI America Supplier No. B48011G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64872-77-1
Molecular Formula C19H18Cl3N3O3S
Molecular Weight (g/mol) 474.781
MDL Number MFCD00058159
InChI Key ZHPWRQIPPNZNML-UHFFFAOYSA-N
Synonym 1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]imidazole Nitrate
PubChem CID 47471
ChEBI CHEBI:3241
IUPAC Name 1-[4-(4-chlorophenyl)-2-(2,6-dichlorophenyl)sulfanylbutyl]imidazole;nitric acid
SMILES C1=CC(=C(C(=C1)Cl)SC(CCC2=CC=C(C=C2)Cl)CN3C=CN=C3)Cl.[N+](=O)(O)[O-]

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 1 g
Formula Weight 474.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butoconazole Nitrate
RTECSNumber NI4399500
TSCA No