missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butenafine Hydrochloride 98.0+%, TCI America™
$225.56 - $665.46
Chemical Identifiers
| CAS | 101827-46-7 |
|---|---|
| Molecular Formula | C23H28ClN |
| Molecular Weight (g/mol) | 353.934 |
| MDL Number | MFCD00917064 |
| InChI Key | LJBSAUIFGPSHCN-UHFFFAOYSA-N |
| PubChem CID | 443867 |
| ChEBI | CHEBI:31325 |
| IUPAC Name | 1-(4-tert-butylphenyl)-N-methyl-N-(naphthalen-1-ylmethyl)methanamine;hydrochloride |
| SMILES | CC(C)(C)C1=CC=C(C=C1)CN(C)CC2=CC=CC3=CC=CC=C32.Cl |
Chemical Identifiers
| 101827-46-7 | |
| 353.934 | |
| LJBSAUIFGPSHCN-UHFFFAOYSA-N | |
| CHEBI:31325 | |
| CC(C)(C)C1=CC=C(C=C1)CN(C)CC2=CC=CC3=CC=CC=C32.Cl |
| C23H28ClN | |
| MFCD00917064 | |
| 443867 | |
| 1-(4-tert-butylphenyl)-N-methyl-N-(naphthalen-1-ylmethyl)methanamine;hydrochloride |
Specifications
| 101827-46-7 | |
| White | |
| MFCD00917064 | |
| LJBSAUIFGPSHCN-UHFFFAOYSA-N | |
| 1-(4-tert-butylphenyl)-N-methyl-N-(naphthalen-1-ylmethyl)methanamine;hydrochloride | |
| 443867 | |
| 353.93 | |
| Crystalline Powder |
| 210°C | |
| C23H28ClN | |
| 1 g | |
| CC(C)(C)C1=CC=C(C=C1)CN(C)CC2=CC=CC3=CC=CC=C32.Cl | |
| 353.934 | |
| CHEBI:31325 | |
| ≥98.0% (HPLC,T) | |
| Butenafine Hydrochloride |
Safety and Handling
RTECSNumber : QJ8585000
TSCA : No