missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butenafine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 210°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 353.93 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Butenafine Hydrochloride |
Safety and Handling
| RTECSNumber | QJ8585000 |
| TSCA | No |