missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butein 98.0+%, TCI America™
$257.24 - $566.67
Chemical Identifiers
| CAS | 487-52-5 |
|---|---|
| Molecular Formula | C15H12O5 |
| Molecular Weight (g/mol) | 272.256 |
| MDL Number | MFCD00017300 |
| InChI Key | AYMYWHCQALZEGT-ORCRQEGFSA-N |
| Synonym | (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′C,3,4,4′C-Tetrahydroxychalcone |
| PubChem CID | 5281222 |
| ChEBI | CHEBI:3237 |
| IUPAC Name | (E)-1-(2,4-dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one |
| SMILES | C1=CC(=C(C=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3803100MG
|
TCI America
B3803100MG |
100 mg |
Each for $257.24
|
|
|||||
|
B38031G
|
TCI America
B38031G |
1 g |
Each for $566.67
|
|
|||||
Chemical Identifiers
| 487-52-5 | |
| 272.256 | |
| AYMYWHCQALZEGT-ORCRQEGFSA-N | |
| 5281222 | |
| (E)-1-(2,4-dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one |
| C15H12O5 | |
| MFCD00017300 | |
| (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′C,3,4,4′C-Tetrahydroxychalcone | |
| CHEBI:3237 | |
| C1=CC(=C(C=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O)O |
Specifications
| 487-52-5 | |
| C15H12O5 | |
| 100 mg | |
| AYMYWHCQALZEGT-ORCRQEGFSA-N | |
| (E)-1-(2,4-dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one | |
| 5281222 | |
| 272.26 | |
| Crystalline Powder |
| 215°C | |
| MFCD00017300 | |
| (E)-1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′C,3,4,4′C-Tetrahydroxychalcone | |
| C1=CC(=C(C=C1C=CC(=O)C2=C(C=C(C=C2)O)O)O)O | |
| 272.256 | |
| CHEBI:3237 | |
| ≥98.0% (HPLC) | |
| Butein |
Safety and Handling
TSCA : No
Recommended Storage : Freezer