accessibility menu, dialog, popup

UserName

CAS RN 487-52-5

OHHOOHOHO(E)

CAS RN 487-52-5

IUPAC Name: (2E)-1-(2,4-dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)prop-2-en-1-one
Synonyms: butein
Molecular Weight (g/mol): 272.26
Molecular Formula: C15H12O5
InChi Key: AYMYWHCQALZEGT-ORCRQEGFSA-N
SMILES: OC1=CC=C(C(=O)\C=C\C2=CC=C(O)C(O)=C2)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results