missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butein, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| CAS | 487-52-5 |
| Linear Formula | C15H12O5 |
| MDL Number | MFCD00017300 |
| Synonym | 1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one; 2 ′,3,4,4 ′-Tetrahydroxychalcone |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 272.25 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |