missing translation for 'onlineSavingsMsg'
Learn More

Butadiene monoxide, 98%

CAS: 930-22-3 | C4H6O | 70.091 g/mol

Supplier:  Thermo Scientific Chemicals L0023806

 View more versions of this product

Catalog No. AAL0023806


Only null left
Explore available promotions

Description

Description

Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents.

Notes
Moisture sensitive. Store in cool place. Incompatible with acids, bases and oxidizing agents.
Specifications

Specifications

Butadiene monoxide
-135°C
65°C to 66°C
Recognizable
1.418
5 g
103170
butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
GXBYFVGCMPJVJX-UHFFFAOYSA-N
2-ethenyloxirane
13586
98%
930-22-3
0.87
−50°C (−58°F)
C4H6O
MFCD00005149
UN1993
Moisture sensitive
Miscible with ethanol,ethyl ether,benzene and organic solvents.
C=CC1CO1
70.091
70.09

Chemical Identifiers

930-22-3
70.091
GXBYFVGCMPJVJX-UHFFFAOYSA-N
13586
C=CC1CO1
C4H6O
MFCD00005149
butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
2-ethenyloxirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H341
Highly flammable liquid and vapor.
Suspected of causing genetic defects.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c

H225-H302-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 213-210-4

RTECSNumber : EM7350000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only