accessibility menu, dialog, popup

UserName

CAS RN 930-22-3

IUPAC Name: 2-ethenyloxirane
Synonyms: oxirane, ethenyl-
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: GXBYFVGCMPJVJX-UHFFFAOYNA-N
SMILES: C=CC1CO1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)