missing translation for 'onlineSavingsMsg'
Learn More

Butadiene monoxide, 98%

Catalog No. AAL0023803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0023803 1 g
AAL0023806 5 g
AAL0023814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0023803 Supplier Thermo Scientific Chemicals Supplier No. L0023803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 930-22-3 | C4H6O | 70.091 g/mol

Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butadiene monoxide is used as key starting material for the synthesis of (-)-bulgecinine and (+)-broussonetine G.

Solubility
Miscible with ethanol, ethyl ether, benzene and organic solvents.

Notes
Moisture sensitive. Store in cool place. Incompatible with acids, bases and oxidizing agents.

Chemical Identifiers

CAS 930-22-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00005149
InChI Key GXBYFVGCMPJVJX-UHFFFAOYSA-N
Synonym butadiene monoxide, oxirane, ethenyl, 3,4-epoxy-1-butene, 2-vinyloxirane, butadiene epoxide, 1,2-epoxy-3-butene, vinyloxirane, butadiene oxide, butadiene monoepoxide, butadiene monooxide
PubChem CID 13586
IUPAC Name 2-ethenyloxirane
SMILES C=CC1CO1

Specifications

Melting Point -135°C
Density 0.87
Boiling Point 65°C to 66°C
Flash Point −50°C (−58°F)
Odor Recognizable
Refractive Index 1.418
Quantity 1 g
UN Number UN1993
Beilstein 103170
Sensitivity Moisture sensitive
Solubility Information Miscible with ethanol,ethyl ether,benzene and organic solvents.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Butadiene monoxide
Show More Show Less
Hazard Category H225-H302-H319
Hazard Statement GHS H Statement
H225-H341
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-210-4
RTECSNumber EM7350000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only