missing translation for 'onlineSavingsMsg'
Learn More

Buspirone hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018335
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018335 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018335 Supplier Merck Emd Millipore Supplier No. PHR1917500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 33386-08-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H31N5O2 · HCl
MDL Number MFCD00078569
Synonym Buspirone hydrochloride; N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5 ]decane-7,9-dione hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 421.96
Grade Certified Reference Material