Learn More
Eisco™ Burette Clamps
Constructed of cast sheet metal, these burette clamps have a built-in boss head for easy attachment to a rod stand
$36.71
Specifications
| For Use With (Equipment) | Burette |
|---|---|
| Material | Cast Metal |
| For Use With (Application) | Titration |
| No. per Pack | 1 |
| No. per Case | 1 |
Description
- Made from Cast Sheet Metal
- Includes built-in boss head with screw adjustment
- Easily attaches to a rod stand
- Single, and Double Burette Clamps are available
Specifications
| Burette | |
| Titration | |
| 1 |
| Cast Metal | |
| 1 |