Learn More
Eisco™ Burette Clamps
Catalog No. S27033
Change view
Click to view available options
Type:
Double Buret Clamp
Catalog No. Type
S27033 Double Buret Clamp
1 options
Catalog No. S27033 Supplier Eisco™ Supplier No. CH0256B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Constructed of cast sheet metal, these burette clamps have a built-in boss head for easy attachment to a rod stand
  • Made from Cast Sheet Metal
  • Includes built-in boss head with screw adjustment
  • Easily attaches to a rod stand
  • Single, and Double Burette Clamps are available
Specifications
For Use With (Equipment) Burette
Material Cast Metal
For Use With (Application) Titration
No. per Pack 1
No. per Case 1
Includes Boss Head
Type Double Buret Clamp