missing translation for 'onlineSavingsMsg'
Learn More

Bupropion Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013910
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013910 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013910 Supplier Merck Emd Millipore Supplier No. PHR17301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bupropion Hydrochloride is an aminoketone derivative and an antidepressant drug, widely used to treat depression. It is also employed for the treatment of obesity, smoking cessation and attention deficit hyperactivity disorder.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 31677-93-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H18 ClNO · HCl
MDL Number MFCD00055209
Synonym Bupropion hydrochloride; (+/-)-1-(3-Chlorophenyl)-2-[(1,1-dimethylethyl)amino]-1-propanone hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 276.20
Grade Certified Reference Material