missing translation for 'onlineSavingsMsg'
Learn More

Buphedrone hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™

Catalog No. B0471ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0471ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0471ML Supplier MilliporeSigma Supplier No. B0471ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Buphedrone is a new synthetic cathinone marketed as 'bath salts' or 'plant food' over the internet.
  • Bath salt products are sold in powder or crystalline form under names including Ivory Wave, Vanilla Sky, Pixie Dust, and White Dove.
  • According to the US Drug Enforcement Administration (DEA), bath salt cathinones have grown increasingly popular among teenagers and young adults due to their ready availability over the internet and legal status.

Specifications

Chemical Name or Material Buphedrone HCl
Packaging Ampule
CAS 166593-10-8
Molecular Formula C11H16ClNO
Concentration 1.0 mg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 213.7
Quantity 1 mL
Grade Certified
Show More Show Less