Learn More
Bumetanide, 98+%
Inhibitor of the sodium-potassium-chloride cotransporter | CAS: 28395-03-1 | C17H20N2O5S | 364.416 g/mol
Supplier: Thermo Scientific Chemicals J6230206
Description
Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1
Solubility
Soluble in ethanol (10 mg/ml), DMSO (25 mg/ml), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.
Specifications
| Bumetanide | |
| 230°C to 231°C | |
| C17H20N2O5S | |
| 5 g | |
| bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum | |
| MAEIEVLCKWDQJH-UHFFFAOYSA-N | |
| 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid | |
| 2471 | |
| 364.4 | |
| Solid |
| 28395-03-1 | |
| White | |
| MFCD00078949 | |
| 14,1484 | |
| Soluble in ethanol (10mg/ml),DMSO (25mg/ml),acetone,benzene,methanol,propylene glycol,and water (<1mg/ml). | |
| CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2 | |
| 364.416 | |
| CHEBI:3213 | |
| ≥98% |
Chemical Identifiers
| 28395-03-1 | |
| 364.416 | |
| MAEIEVLCKWDQJH-UHFFFAOYSA-N | |
| 2471 | |
| 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid |
| C17H20N2O5S | |
| MFCD00078949 | |
| bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum | |
| CHEBI:3213 | |
| CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2 |
Safety and Handling
EINECSNumber : 249-004-6
RTECSNumber : DG4910000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only