missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Excel FR™ Lab Coats
Protection: Arc Rating ATPV 7.7 calories/cm2
Supplier: Bulwark™ KEL2LBRGXL
Description
Protection: Arc Rating ATPV 7.7 calories/cm2
- Flame-resistant, 7 oz. 100% cotton
- Button-front closure with lapel collar
- Hemmed sleeve ends
- Side-vent openings
- Light blue
- Hazard rating class 1
Specifications
| Flame-resistant Lab Coat | |
| Unisex | |
| Light Blue | |
| Hemmed | |
| Non-sterile | |
| 3 | |
| KEL2LBRG | |
| 121.92 to 127 cm | |
| 7.7 Calories/cm2 | |
| Chemical, Oil, Gas, Petrochemical, Electrical, Utilities | |
| 42.5 in. | |
| 7 oz. |
| X-Large | |
| Cotton | |
| Buttons | |
| Reusable | |
| Flame | |
| Lapel | |
| 48 to 50 in. | |
| Class 1 | |
| ASTM F2302, NFPA 70E, ASTM F1506 | |
| Side Vent Openings | |
| 107.95 cm | |
| 0.19 kg |