missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Excel FR™ Lab Coats
Description
Protection: Arc Rating ATPV 7.7 calories/cm2
- Flame-resistant, 7 oz. 100% cotton
- Button-front closure with lapel collar
- Hemmed sleeve ends
- Side-vent openings
- Light blue
- Hazard rating class 1
Specifications
Specifications
| Product Type | Flame-resistant Lab Coat |
| Size | Medium |
| Gender | Unisex |
| Material | Cotton |
| Color | Light Blue |
| Closure Type | Buttons |
| Cuff Style | Hemmed |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Flame |
| Show More |