missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Industrial Breathing Air Supply Hoses

For use with breathing air compressors or breathing air cylinders.

$752.53 - $752.53

Specifications

Color Red
Product Type Air Supply Hose
Diameter (English) Interior 0.37 in.
Diameter (Metric) Interior 0.93 cm
View More Specs

Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
18-998-074
View Documents
Bullard™
V55030
Each for $752.53
Only null left
 
Description

Description

  • Coiled design assists in workplace cleanliness
  • Nylon construction
  • 3/8 in. I.D.
  • Choose from industrial interchange or Schrader quick-disconnect coupler model

Specifications

Specifications

Red
Air Supply Hose
0.37 in.
0.93 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'