missing translation for 'onlineSavingsMsg'
Learn More

Bullard™ Industrial Breathing Air Supply Hoses

Catalog No. 18998074
Catalog No. 18998074 Supplier Bullard™ Supplier No. V55030

For use with breathing air compressors or breathing air cylinders.

  • Coiled design assists in workplace cleanliness
  • Nylon construction
  • 3/8 in. I.D.
  • Choose from industrial interchange or Schrader quick-disconnect coupler model

Specifications

Color Red
Product Type Air Supply Hose
Model V55030
Diameter (English) Interior 0.37 in.
Diameter (Metric) Interior 0.93 cm