Learn More
Thermo Scientific™ Bulk Pack HDPE Cylinder Rounds without Caps
GREENER_CHOICE
Use the HDPE cylinder round as a general purpose container. Sold in bulk, unassembled and unprocessed, these containers are ready for your own cleaning procedure and assembly with caps purchased separately.
$410.27 - $679.33
Specifications
Certification Parameters None
Color Natural
Material HDPE
Sterility Non-sterile
Transparency Translucent
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity & Availability  
05712214
GREENER_CHOICE
Accessories View Documents Promotion Details
Thermo Scientific™
013 0250BP
250 mL 24-410 mm
Case of 230 Each for $679.33
Only null left
05712225
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
013 0500BP
500 mL 28-410 mm
Case of 120 Each for $418.24
Only null left
05712224
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
013 1000BP
1000 mL 28-410 mm
Case of 65 Each for $410.27
Only null left
Description
Description
Thermo Scientific™ HDPE Cylinder Round is a general purpose container. Sold in bulk, unassembled and unprocessed, these containers are ready for your own cleaning procedure and assembly with caps purchased separately. The same high quality container component as our assembled HDPE cylinder rounds, but sold in bulk, without a cap. Pair them with a matching bulk cap (Cat. 067-series) sold separately or another cap of the same finish. Unprocessed and uncertified.
  • HDPE container, no cap
  • Caps sold separately: 067-2410WBP or 067-2810WBP
  • Narrow mouth for precision pouring
  • Slim bottle design for easy grip and handling
  • Recommended for general purpose sampling where cleaning and certification is not required, or ready for your own cleaning and certification procedures
  • Economically bulk packed without caps
Specifications
Specifications
None
HDPE
Translucent
N/A
None
Natural
Non-sterile
None
Unprocessed
Round