missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poltex Bulk Cone Mask Dispenser
Clear dispenser holds a box or stack of cone masks. Poltex Blue Cone Mask Dispenser is has a hinged lid to keep dust out.
$158.21 - $198.50
Specifications
| Color | Clear |
|---|---|
| Material | PETG |
| For Use With (Application) | Storing and dispensing cone masks |
| Dimensions (D x W x H) Interior | 5.5 x 5.5 x 7 in. |
Description
Clear dispenser holds a box or stack of cone masks. Poltex Blue Cone Mask Dispenser is has a hinged lid to keep dust out.
- Holds a stack of masks or the whole box.
- Made from durable 3/16 in. thick PETG
- Hinged lid keeps dust out
- Interior DxWxH: 5.5 x 5.5 x 7 in.
- Clear
- Available with magnets or holes for mounting
Specifications
| Clear | |
| PETG | |
| Storing and dispensing cone masks | |
| 5.5 x 5.5 x 7 in. |