missing translation for 'onlineSavingsMsg'
Learn More

Poltex Bulk Cone Mask Dispenser

Catalog No. 22000232
Encompass
Change view
Click to view available options
Mounting:
Magnetic Mount
Pre-drilled Holes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22000232 Magnetic Mount
22000231 Pre-drilled Holes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000232 Supplier Poltex Supplier No. CONMSKM
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear dispenser holds a box or stack of cone masks. Poltex Blue Cone Mask Dispenser is has a hinged lid to keep dust out.

  • Holds a stack of masks or the whole box.
  • Made from durable 3/16 in. thick PETG
  • Hinged lid keeps dust out
  • Interior DxWxH: 5.5 x 5.5 x 7 in.
  • Clear

  • Available with magnets or holes for mounting

Specifications

Color Clear
Material PETG
For Use With (Application) Storing and dispensing cone masks
Dimensions (D x W x H) Interior 5.5 x 5.5 x 7 in.
Mounting Magnetic Mount