Learn More
Bulbtronics™ Fluorescent Lamps DFS Item
Supplier:  Bulbtronics™ 0015696
 View more versions of this product
Catalog No. 14-395-888
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Medium Bi-Pin (G13)
40 W
Linear Fluorescent Lamp with 6 mm Reflective Strip
3.81 cm
121.9 cm
Fluorescent
Blacklight Blue
1.5 in.
48 in.
T12
Product Suggestions
Product Suggestions