missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BugBuster™ Protein Extraction Reagent

Catalog No. 705844
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
705844 500 mL
705843 100 mL
705841000ML 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 705844 Supplier MilliporeSigma™ Supplier No. MIL705844

Reagent for protein sample preparation

  • Formulated for gentle disruption of cell wall of E. coli, resulting in liberation of soluble protein
  • Provides simple, rapid, low cost alternative to mechanical methods
  • Formulation utilizes mixture of non-ionic detergents capable of cell wall perforation without denaturing soluble protein

Specifications

Quantity 500 mL
Content And Storage Store at 15°C to 30°C