missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BugBuster™ (primary amine-free) Extraction Reagent

Catalog No. 709234
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709234 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 709234 Supplier MilliporeSigma™ Supplier No. 709234

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideally suited for extraction of metal-dependent proteins

  • Special formulation of BugBuster designed for applications where primary amines would interfere if present in protein extract, such as protein immobilization or cross-linking
  • The PIPPS buffer used in BugBuster (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins

Specifications

Quantity 500mL
Content And Storage Store at+15°C to+30°C