missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BugBuster™ HT Protein Extraction Reagent

Catalog No. 709223
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709223 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 709223 Supplier MilliporeSigma™ Supplier No. 709223

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eliminates common bioprocessing problems resulting from traditional cell lysis procedures

  • Combines BugBuster Protein Extraction Reagent and Benzonase™ Nuclease in one convenient reagent
  • Soluble proteins are gently extracted from E. coli without exposure to heat or oxidative damage
  • Viscosity is eliminated by nucleic acid digestion in a single step
  • The resulting protein extract can easily be fractionated by conventional purification techniques
  • Ideally suited for application in high throughput protein purifications

Specifications

Quantity 100mL
Content And Storage Store at +15° to +30°C