missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BugBuster™ 10X Protein Extraction Reagent

Catalog No. 709215
Change view
Click to view available options
Quantity:
10mL
50mL
100mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709215 100mL
709214 50mL
709213 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 709215 Supplier MilliporeSigma™ Supplier No. 709215

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent for protein sample preparation

  • Concentrated formulation of proprietary detergents without addition of salts or buffer components
  • Provides flexible alternative to ready-to-use standard 1X BugBuster, allowing user-defined dilution and addition of buffer components
  • Consists all of the bioprocessing benefits of standard BugBuster plus the freedom to control pH, reagent concentration, and buffer additives necessary for maximum extraction and activity of your target protein

Specifications

Quantity 100mL
Content And Storage Store at+15°C to+30°C