missing translation for 'onlineSavingsMsg'
Learn More

Buflomedil Hydrochloride 98.0+%, TCI America™

Catalog No. p-7123072
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B36471G 1 g
B36475G 5 g
2 options

Catalog No. B36471G

Supplier: TCI America B36471G

Only null left

Chemical Identifiers

CAS 35543-24-9
Molecular Formula C17H26ClNO4
Molecular Weight (g/mol) 343.848
MDL Number MFCD00078965
InChI Key ZDPACSAHMZADFZ-UHFFFAOYSA-N
Synonym 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone Hydrochloride
PubChem CID 6420013
IUPAC Name 4-pyrrolidin-1-yl-1-(2,4,6-trimethoxyphenyl)butan-1-one;hydrochloride
SMILES COC1=CC(=C(C(=C1)OC)C(=O)CCCN2CCCC2)OC.Cl

Specifications

Melting Point 193°C
Color White
Quantity 1 g
Formula Weight 343.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Buflomedil Hydrochloride
RTECSNumber EL9900000
TSCA No